missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Pluripotent Stem Cell-Derived Endoderm qPCR Array
Description
- The Human Pluripotent Stem Cell (hPSC)-Derived Endoderm Quantitative Polymerase Chain Reaction (qPCR) Array is designed for characterization of hPSC-derived definitive endodermal progenitor cells and their differentiated progeny including pancreatic, hepatic, and intestinal cells
- This qPCR array provides the gene expression profile of hPSC-derived endodermal cells following in vitro differentiation, and can aid in development of protocols for directed differentiation towards endodermal lineages
- Genes were selected based on their demonstrated expression in hPSCs, or in hPSC-derived ectodermal, mesodermal, and endodermal lineage cells (D'Amour et al, including pancreatic (Kroon et al, hepatic (Hay et al, and intestinal progenitor cells (Grapin-Botton & Melton)
Specifications
Specifications
| Description | Endoderm, PSC-Derived; Hepatic Cells; Intestinal Cells; Pancreatic Cells; Pluripotent Stem Cells |
| Format | 96-well Plate Type 4 |
| For Use With (Application) | Real Time PCR (qPCR) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?