missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Pluripotent Stem Cell-Derived Endoderm qPCR Array

Product Code. 17149291
Encompass_Preferred
Change view
Click to view available options
Format:
96-well Plate Type 1
96-well Plate Type 3
96-well Plate Type 4
Unit Size:
1 set
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format unitSize
17149291 96-well Plate Type 1 1 set
17159291 96-well Plate Type 3 1 set
17169291 96-well Plate Type 4 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17149291 Supplier STEMCELL Technologies Supplier No. 07531

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For characterization of hPSC-derived progenitor cells of definitive endoderm, hepatic, intestinal, or pancreatic lineages

  • The Human Pluripotent Stem Cell (hPSC)-Derived Endoderm Quantitative Polymerase Chain Reaction (qPCR) Array is designed for characterization of hPSC-derived definitive endodermal progenitor cells and their differentiated progeny including pancreatic, hepatic, and intestinal cells
  • This qPCR array provides the gene expression profile of hPSC-derived endodermal cells following in vitro differentiation, and can aid in development of protocols for directed differentiation towards endodermal lineages
  • Genes were selected based on their demonstrated expression in hPSCs, or in hPSC-derived ectodermal, mesodermal, and endodermal lineage cells (D'Amour et al, including pancreatic (Kroon et al, hepatic (Hay et al, and intestinal progenitor cells (Grapin-Botton & Melton)

Specifications

Description Endoderm, PSC-Derived; Hepatic Cells; Intestinal Cells; Pancreatic Cells; Pluripotent Stem Cells
Format 96-well Plate Type 1
For Use With (Application) Real Time PCR (qPCR)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.