missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PLLP (aa 1-35) Control Fragment Recombinant Protein

Product Code. 30180533
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30180533

Brand: Invitrogen™ RP98551

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Plasmolipin is an 18 kDa membrane-bound proteolipid that belongs to the tetraspan (4TM) family, a rapidly growing group of myelin-associated proteins. Many of these proteins could be linked to human hereditary demyelinating neuropathies. Studies indicate that plasmolipin is a component of the cytoskeletal structure and has been localized to the apical surface of tubular epithelial cells. It has been shown that, upon addition to lipid bilayers, plasmolipin induces formation of ion channels that are both voltage-dependent and potassium-specific. Plasmolipin was originally isolated from kidney tissue. Subsequently, it has been shown to be widely expressed, not only being present in kidney, but also in the nervous system, thymus, testis, lung, and thyroid gland.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y342
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51090
Name Human PLLP (aa 1-35) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 0610010I06Rik; AV001002; Plapi; Plasma membrane proteolipid; plasma membrane proteolipid (plasmolipin); plasmolipin; PLLP; PMLP; Tm4sf11; transmembrane 4 superfamily member 11; transmembrane 4 superfamily member 11 (plasmolipin)
Common Name PLLP
Gene Symbol PLLP
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MAEFPSKVSTRTSSPAQGAEASVSALRPDLGFVRS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.