missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PlGF Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems BAF264
This item is not returnable.
View return policy
Description
PLGF Polyclonal specifically detects PLGF in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| PLGF | |
| Polyclonal | |
| Biotin | |
| CAA38698 | |
| PGF | |
| E. coli-derived recombinant human PlGF (R&D Systems, Catalog # 264-PG) Ala21-Arg149 Accession # CAA38698 | |
| 50 μg | |
| Primary | |
| Detects human PlGF in ELISAs and Western blots. In sandwich immunoassays, less than 20% cross-reactivity with recombinant human (rh) VEGF/PlGF is observed and less than 0.07% cross-reactivity with rhFlt-1, recombinant mouse PlGF-2, rhPDGF, and rhVEGF is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| D12S1900, PGF, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5228 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction