missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PlGF Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2643-100
This item is not returnable.
View return policy
Description
PLGF Monoclonal antibody specifically detects PLGF in Human samples. It is validated for Immunocytochemistry
Specifications
| PLGF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| D12S1900, PGF, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760 | |
| E. coli-derived human PlGF, Ala21-Arg149, Accession # P49763-2 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Immunocytochemistry | |
| 1038864 | |
| Immunocytochemistry 8-25 ug/mL | |
| P49763-2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5228 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction