missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human PlGF-1, Animal-Free Recombinant Protein, PeproTech®

Product Code. 17896548
Change view
Click to view available options
Quantity:
250 μg
5 μg
Unit Size:
250µg
5µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17896548 5 μg 5µg
17836478 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17896548 Supplier Gibco™ Supplier No. AF100065UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human PlGF-1 is a 29.7 kDa disulfide-linked homodimeric protein of two 132 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

PlGF-1 is an angiogenic factor that belongs to the cysteine-knot superfamily of growth factors. PlGF-1 is expressed in placental tissues, the colon, and mammary carcinomas. It signals through the VEGFR-1/FLT1 receptor, and stimulates endothelial cell proliferation and migration.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P49763
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 5228
Molecular Weight (g/mol) 29.7 kDa
Name Human PlGF-1, Animal-Free
Purification Method HPLC, SDS-PAGE
Quantity 5 μg
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias Placenta Growth Factor; Placenta Growth Factor-1
Common Name PlGF-1
Gene Symbol PGF
Biological Activity Determined by its ability to chemoattract human monocytes of using a concentration range of 1.0-10.0 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MLPAVPPQQW ALSAGNGSSE VEVVPFQEVW GRSYCRALER LVDVVSEYPS EVEHMFSPSC VSLLRCTGCC GDENLHCVPV ETANVTMQLL KIRSGDRPSY VELTFSQHVR CECRPLREKM KPERCGDAVP RR
Content And Storage -20°C
Activity Determined by its ability to chemoattract human monocytes of using a concentration range of 1.0-10.0 ng/ml
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.