Learn More
Abnova™ Human PLG native Protein from human plasma
Description
Plasminogen (PLG) is a circulating zymogen that is converted to the active enzyme plasmin by cleavage of the peptide bond between arg560 and val561, which is mediated by urokinase (PLAU; MIM 191840) and tissue plasminogen activator (PLAT; MIM 173370). The main function of plasmin is to dissolve fibrin (see, e.g., FGA, MIM 134820) clots. Plasmin, like trypsin, belongs to the family of serine proteinases (Miyata et al., 1982 [PubMed 6216475]; Forsgren et al., 1987 [PubMed 3030813]).[supplied by OMIM]
Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | In 200mM Sodium Phosphate, 1mM 6-aminohexanoic acid, pH 7.3 (50% glycerol). |
| Gene ID (Entrez) | 5340 |
| Molecular Weight (g/mol) | 81kDa |
| Name | PLG (Human) Native Protein |
| Preparation Method | Native protein purified from human plasma. |
| Quantity | 1 mg |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Regulatory Status | RUO |
| Gene Alias | DKFZp779M0222 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.