missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PLG native Protein from human plasma

Product Code. 16068096
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16068096 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16068096 Supplier Abnova™ Supplier No. P4958.1mg
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for Func, SDS-PAGE

Plasminogen (PLG) is a circulating zymogen that is converted to the active enzyme plasmin by cleavage of the peptide bond between arg560 and val561, which is mediated by urokinase (PLAU; MIM 191840) and tissue plasminogen activator (PLAT; MIM 173370). The main function of plasmin is to dissolve fibrin (see, e.g., FGA, MIM 134820) clots. Plasmin, like trypsin, belongs to the family of serine proteinases (Miyata et al., 1982 [PubMed 6216475]; Forsgren et al., 1987 [PubMed 3030813]).[supplied by OMIM]

Specifications

Concentration 4.1 mg/mL
For Use With (Application) Functional Study, SDS-PAGE
Formulation In 0.1M Hepes, 0.1M NaCl, pH 7.4
Gene ID (Entrez) 5340
Molecular Weight (g/mol) 92kDa
Name PLG (Human) Native Protein
Preparation Method Native protein purified from human plasma
Purification Method Lysine sepharose chromatography
Quantity 1 mg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp779M0222
Common Name PLG
Gene Symbol PLG
Biological Activity No plasmin activity is detected with chromogenic substrate S-2251.
> 98% conversion to plasmin is observed upon activation with uPA.
Species Human
Protein Tag None
Expression System Native protein purified from human plasma
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.