missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PLEKHG6 Control Fragment Recombinant Protein

Product Code. 30194188
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30194188

Brand: Invitrogen™ RP106834

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (77%), Rat (77%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66151 (PA5-66151. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PLEKHG6 functions as a guanine nucleotide exchange factor activating the small GTPase RHOA, which, in turn, induces myosin filament formation. It activates RHOG but not activate RAC1, or to a much lower extent than RHOA and RHOG. Part of a functional unit, involving MyoGEF, MYH10 and RHOA, at the cleavage furrow to advance furrow ingression during cytokinesis. In epithelial cells, required for the formation of microvilli and membrane ruffles on the apical pole. Along with EZR, required for normal macropinocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q3KR16
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55200
Name Human PLEKHG6 Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias BC026778; Gm155; MyoGEF; myosin II interacting GEF; myosin interacting guanine nucleotide exchange factor; myosin-interacting guanine nucleotide exchange factor; PH domain-containing family G member 6; pleckstrin homology and RhoGEF domain containing G6; pleckstrin homology domain containing, family G (with RhoGef domain) member 6; pleckstrin homology domain-containing family G member 6; PLEKHG6
Common Name PLEKHG6
Gene Symbol PLEKHG6
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KAFGPPHEGPLQGLVASRIETYGGRHRASAQSTTGRLYPRGYPVLDPSRRRLQQYVPFARGSGQARGLSPMRLRDPEPEKR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.