missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PLCL2 (aa 1064-1127) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-140102 (PA5-140102. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May play an role in the regulation of Ins(1,4,5)P3 around the endoplasmic reticulum.
Specifications
Specifications
| Accession Number | Q9UPR0 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 23228 |
| Name | Human PLCL2 (aa 1064-1127) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | inactive phospholipase C-like protein 2; KIAA1092; mKIAA1092; phospholipase C; phospholipase C like 2; phospholipase C, epsilon 2; phospholipase C-epsilon-2; phospholipase C-L2; phospholipase C-like 2; PLCE2; PLC-epsilon-2; PLC-L(2); Plcl2; PLC-L2; PRIP-2 |
| Common Name | PLCL2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction