missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Plasminogen ELISA Kit (Chemiluminescence)

Product Code. 18605420 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18605420 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18605420 Supplier Novus Biologicals™ Supplier No. NBP268098

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Chemiluminescence)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 125.00 to 8000 pg/mL
Assay Sensitivity 75.00 pg/mL
Product Type ELISA Kit (Chemiluminescence)
Sample Type Serum, Plasma, Biological Fluids
Specificity This kit recognizes Human Plg in samples. No significant cross-reactivity or interference between Human Plg and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 5340
Gene Symbol PLG
Quantity 1 Kit
Research Discipline Angiogenesis, Cancer, Cardiovascular Biology, Cell Cycle and Replication
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym DKFZp779M0222, EC 3.4.21, EC 3.4.21.7, plasmin, plasminogen
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.