missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PLA2G2E (aa 19-49) Control Fragment Recombinant Protein

Product Code. 30196614
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30196614

Brand: Invitrogen™ RP103543

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64150 (PA5-64150. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Secretory calcium-dependent phospholipase A2 that primarily targets extracellular phospholipids (PubMed:10681567, PubMed:11922621, PubMed:28883454). Hydrolyzes the ester bond of the fatty acyl group attached at sn-2 position of phospholipids (phospholipase A2 activity), releasing various unsaturated fatty acids including oleoate, linoleoate, arachidonate, docosahexaenoate and lysophosphatidylethanolamines in preference to lysophosphatidylcholines (PubMed:10681567, PubMed:28883454). In response to high-fat diet, hydrolyzes minor lipoprotein phospholipids including phosphatidylserines, phosphatidylinositols and phosphatidylglycerols, altering lipoprotein composition and fat storage in adipose tissue and liver. May act in an autocrine and paracrine manner (PubMed:11922621). Contributes to lipid remodeling of cellular membranes and generation of lipid mediators involved in pathogen clearance. Cleaves sn-2 fatty acyl chains of phosphatidylglycerols and phosphatidylethanolamines, which are major components of membrane phospholipids in bacteria (PubMed:11922621). Acts as a hair follicle phospholipase A2. Selectively releases lysophosphatidylethanolamines (LPE) and various unsaturated fatty acids in skin to regulate hair follicle homeostasis. May regulate the inflammatory response by releasing arachidonate, a precursor of prostaglandins and leukotrienes (PubMed:11922621). Upon allergen exposure, may participate in allergic inflammatory response by enhancing leukotriene C4 synthesis and degranulation in mast cells. [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NZK7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 30814
Name Human PLA2G2E (aa 19-49) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias GIIE sPLA2; Group IIE secretory phospholipase A2; mGIIEsPLA2s; phosphatidylcholine 2-acylhydrolase 2 E; phospholipase A2 group IIE; phospholipase A2, group IIE; Pla2g2e; sPLA2-IIE
Common Name PLA2G2E
Gene Symbol PLA2G2E
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GNLVQFGVMIEKMTGKSALQYNDYGCYCGIG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.