missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PKR Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB1980
This item is not returnable.
View return policy
Description
PKR Monoclonal specifically detects PKR in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Immunocytochemistry.
Specifications
| PKR | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunoprecipitation, Immunocytochemistry 8-25 ug/mL | |
| double stranded RNA activated protein kinase, EC 2.7.11.1, eIF-2A protein kinase 2, EIF2AK1, EIF2AK2, eukaryotic translation initiation factor 2-alpha kinase 2P1/eIF-2A protein kinase, interferon-induced, double-stranded RNA-activated protein kinase, interferon-inducible double stranded RNA dependent, interferon-inducible elF2alpha kinase, Interferon-inducible RNA-dependent protein kinase, P68 Kinase, PKRp68 kinase, PPP1R83, PRKR, PRKRMGC126524, Protein Kinase R, Protein kinase RNA-activated | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5610 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunoprecipitation, Immunocytochemistry | |
| HL71/10 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PRKRA | |
| Protein kinase fraction from interferon-treated Daudi human Burkitt's lymphoma cell line. Laurent, A.G. et al. (1985) Proc. Natl. Acad. Sci. 82:4341. | |
| 100 μg | |
| Primary | |
| Detects human PKR in Western blots. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction