missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PKM2 Recombinant Protein

Product Code. 11589051
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μg
missing translation for 'unitSize'
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11589051 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11589051 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' RP75743

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This product contains purified recombinant full length human PKM2 Pyruvate Kinase protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

PKM2 is a pyruvate kinase that catalyzes the production of phosphoenolpyruvate from pyruvate and ATP. PKM2 has been shown to interact with thyroid hormone, and thus may mediate cellular metabolic effects induced by thyroid hormones. PKM2 has been found to bind Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, suggesting a role of this protein in bacterial pathogenesis. Three alternatively spliced transcript variants encoding two distinct isoforms of PKM2 have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14618
Concentration 0.295 mg/mL
For Use With (Application) Control, Western Blot
Formulation PBS with 50% glycerol and no preservative; pH 7.4
Gene ID (Entrez) 5315
Name Human PKM2
pH Range 7.4
Quantity 100 μg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias AA414905; AL024370; AL024424; ATP:pyruvate 2-o-phosphotransferase; CTHBP; Cytosolic thyroid hormone-binding protein; epididymis secretory protein Li 30; HEL-S-30; isozymes M1/M2; m2pk; muscle pyruvate kinase; OIP3; OIP-3; OPA-interacting protein 3; p58; Pk., muscle type; PK2; Pk.-2; Pk3; Pk.-3; PKLR; PKM; pkm.L; PKM12; PKM2; Pykm; pyruvate kinase; pyruvate kinase 2/3; pyruvate kinase 3; pyruvate kinase isozymes M1/M2; pyruvate kinase M1/2; pyruvate kinase M1/2 L homeolog; pyruvate kinase M1/2 L homeolog; pyruvate kinase, muscle; pyruvate kinase muscle isozyme; Pyruvate kinase PKM; pyruvate kinase, liver and RBC; pyruvate kinase, muscle; TCB; THBP1; thyroid hormone-binding protein 1; thyroid hormone-binding protein, cytosolic; tu; tumor M2-Pk.; XELAEV_18018020mg
Common Name PKM2
Gene Symbol PKM
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Met1-Pro531 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.