missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PKC epsilon Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5134-SP
This item is not returnable.
View return policy
Description
PKC epsilon Monoclonal specifically detects PKC epsilon in Human samples. It is validated for Immunohistochemistry.
Specifications
| PKC epsilon | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q02156 | |
| PRKCE | |
| E. coli-derived recombinant human PKC epsilon Gln580-Pro737 Accession # Q02156 | |
| 25 μg | |
| Primary | |
| Detects human PKC epsilon in direct ELISAs. In direct ELISAs, less than 20% cross-reactivity with recombinant human (rh) PKC delta and rhPKC iota is observed. No cross-reactivity with rhPKC alpha , beta 1, beta 2, gamma , eta , μ, nu , theta , or zeta is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 666823 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.11, EC 2.7.11.13, MGC125656, nPKC-epsilon, PKCE, PKCEMGC125657, PRKCE, protein kinase C epsilon type, protein kinase C, epsilon | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5581 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction