missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PIWIL1/HIWI Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6548-SP
This item is not returnable.
View return policy
Description
PIWIL1/HIWI Monoclonal specifically detects PIWIL1/HIWI in Human samples. It is validated for Immunocytochemistry.
Specifications
| PIWIL1/HIWI | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| NP_004755 | |
| PIWIL1 | |
| E. coli-derived recombinant human PIWIL1/HIWI Arg82-Thr290 Accession # NP_004755 | |
| 25 μg | |
| Primary | |
| Detects human PIWIL1/HIWI in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) PIWIL2 or rhPIWIL4 is observed. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 585509 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HIWI, HIWIpiwi homolog, MIWI, PIWI, piwi (Drosophila)-like 1, piwi-like 1 (Drosophila), piwi-like protein 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9271 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido