missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PITX2 Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF7388
This item is not returnable.
View return policy
Description
PITX2 Polyclonal specifically detects PITX2 in Human samples. It is validated for Immunohistochemistry.
Specifications
| PITX2 | |
| Polyclonal | |
| Unconjugated | |
| Q99697 | |
| PITX2 | |
| E. coli-derived recombinant human PITX2 Ala149-Val317 Accession # Q99697 | |
| 100 μg | |
| Primary | |
| Detects human PITX2 in direct ELISAs. In direct ELISAs, less than 5% cross-reactivity with recombinant human PITX1 is observed. | |
| Human | |
| IgG |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL | |
| all1-responsive gene 1, ALL1-responsive protein ARP1, ARP1MGC20144, Brx1, Homeobox protein PITX2, IDG2, IGDS, IGDS2, IHG2, IRID2, Otlx2, paired-like homeodomain 2, Paired-like homeodomain transcription factor 2MGC111022, pituitary homeo box 2, pituitary homeobox 2, PTX2, RGS, RGSRIEG1Brx1, RIEG, RIEG bicoid-related homeobox transcription factor, rieg bicoid-related homeobox transcription factor 1, RIEG1, RP1, RS, solurshin | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 5308 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction