missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PIT1 (aa 184-249) Control Fragment Recombinant Protein

Product Code. 30182826
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182826

Brand: Invitrogen™ RP98326

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59662 (PA5-59662. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PIT1 is a pituitary-specific transcription factor responsible for pituitary development and hormone expression in mammals and is a member of the POU family of transcription factors that regulate mammalian development. The POU family is so named because the first 3 members identified were PIT1 and OCT1 of mammals, and Unc-86 of C. elegans. PIT1 contains 2 protein domains, termed POU-specific and POU-homeo, which are both necessary for high affinity DNA binding on genes encoding growth hormone and prolactin. PIT1 is also important for regulation of the genes encoding prolactin and thyroid-stimulating hormone beta subunit by thyrotropin-releasing hormone and cyclic AMP.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P28069
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5449
Name Human PIT1 (aa 184-249) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CPHD1; dw; dwarf; GHF1; GHF-1; GHF1A; growth hormone factor 1; Hmp1; homeodomain protein for Growth Hormone (GH); PIT1; pit-1; Pit1-rs1; PIT1Z; Pituary specific transcription factor 1 (growth hormone factor 1); pituitary specific transcription factor 1; pituitary specific transcription factor 1, related sequence 1; pituitary-specific positive transcription factor 1; pituitary-specific positive transcription factor 1 variant w; POU class 1 homeobox 1; POU domain, class 1, transcription factor 1; POU domain, class 1, transcription factor 1 (Pit1, growth hormone factor 1); POU1F1; POU1F1a; transcription factor Pit-1
Common Name PIT1
Gene Symbol POU1F1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence CKLKAILSKWLEEAEQVGALYNEKVGANERKRKRRTTISIAAKDALERHFGEQNKPSSQEIMRMAE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado