missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PIP4K2A (aa 317-351) Control Fragment Recombinant Protein

Product Code. 30200304
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200304

Brand: Invitrogen™ RP105356

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PIP4K2A lipid kinase or phosphatidylinositol-5-phosphate 4-kinase, type II, alpha, is a member of the phosphatidylinositol-5-phosphate 4-kinase family that catalyzes the phosphorylation of phosphatidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-4,5-bisphosphate. PIP4K2A plays as essential role in the phosphoinositide signal transduction cascades as the precursor to second messengers and is involved in the regulation of secretion, cell proliferation, differentiation, and motility. PIP4K2A may be one of the factors related to the regulation of the beta-globin gene expression and the different levels of Hb H in alpha-thalassemic patients. Diseases associated with PIP4K2A include B-Lymphoblastic Leukemia/Lymphoma With Hyperdiploidy and Bipolar Disorder.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P48426
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 5305
Name Human PIP4K2A (aa 317-351) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1-phosphatidylinositol 5-phosphate 4-kinase 2-alpha; 1-phosphatidylinositol-4-phosphate kinase; 1-phosphatidylinositol-4-phosphate-5-kinase; 1-phosphatidylinositol-5-phosphate 4-kinase 2-alpha; 1-phosphatidylinositol-5-phosphate 4-kinase A; AW742916; diphosphoinositide kinase 2-alpha; Phosphatidylinositol 5-phosphate 4-kinase type II alpha; phosphatidylinositol 5-phosphate 4-kinase type-2 alpha; phosphatidylinositol-4-phosphate 5-kinase, type II, alpha; phosphatidylinositol-5-phosphate 4-kinase type 2 alpha; phosphatidylinositol-5-phosphate 4-kinase type-2 alpha; phosphatidylinositol-5-phosphate 4-kinase, type II, alpha; PI(5)P 4-kinase type II alpha; PI5P4KA; Pip4k2a; PIP4KII-alpha; PIP5K2; Pip5k2a; PIP5KIIA; PIP5KIIalpha; PIP5KII-alpha; PIP5KIII; PIPK; PIPK2 alpha; PtdIns(4)P-5-kinase B isoform; PtdIns(4)P-5-kinase C isoform; ptdIns(5)P-4-kinase isoform 2-alpha; type II phosphatidylinositol-4-phosphate 5-kinase 53 K isoform
Common Name PIP4K2A
Gene Symbol PIP4K2A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PGNTLNSSPPLAPGEFDPNIDVYGIKCHENSPRKE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.