missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PIEZO2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
£195.00 - £450.00
Specifications
| Antigen | PIEZO2 |
|---|---|
| Clone | 1081503 |
| Dilution | Immunohistochemistry 0.5-15 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30245505
|
R&D Systems
MAB11559-SP |
25 μg |
£195.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30245514
|
R&D Systems
MAB11559-100 |
100 μg |
£450.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PIEZO2 Monoclonal antibody specifically detects PIEZO2 in Human samples. It is validated for ImmunohistochemistrySpecifications
| PIEZO2 | |
| Immunohistochemistry 0.5-15 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| C18orf30, C18orf58, DA3, DA5, DAIPT, FAM38B, FAM38B2, family with sequence similarity 38, member A pseud, family with sequence similarity 38, member B, family with sequence similarity 38, member B2, FLJ23144, FLJ23403, FLJ25916, FLJ34907, FLJ37734, FLJ45725, HsT748, HsT771, MWKS, piezo-type mechanosensitive ion channel component, piezo-type mechanosensitive ion channel component 2, protein PIEZO2 | |
| PIEZO2 containing synthetic peptide, Accession # Q9H5I5 | |
| Primary | |
| Reconstitute lyophilized material at 0.2mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1081503 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9H5I5 | |
| 63895 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title