missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Piccolo Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7935-SP
This item is not returnable.
View return policy
Description
Piccolo Polyclonal specifically detects Piccolo in Human samples. It is validated for Immunohistochemistry.
Specifications
| Piccolo | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACZ, Aczonin, PCLO, piccolo (presynaptic cytomatrix protein), protein piccolo | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 27445 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL | |
| NP_149015 | |
| PCLO | |
| E. coli-derived recombinant human Piccolo Ala4065-Phe4328 Accession # NP_149015 | |
| 25 μg | |
| Primary | |
| Detects human Piccolo in direct ELISAs. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction