missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PIAS2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4768
This item is not returnable.
View return policy
Description
PIAS2 Polyclonal specifically detects PIAS2 in Human samples. It is validated for Western Blot.
Specifications
| PIAS2 | |
| Polyclonal | |
| Unconjugated | |
| O75928 | |
| PIAS2 | |
| E. coli-derived recombinant human PIAS2 Met1-Gly550 Accession # O75928 | |
| 100 μg | |
| Primary | |
| Detects human PIAS2 in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Androgen receptor-interacting protein 3, ARIP3, DAB2-interacting protein, DIP, E3 SUMO-protein ligase PIAS2, MGC102682, miz, Miz1, Msx-interacting zinc finger protein, Msx-interacting-zinc finger, PIAS-NY protein, PIASX, PIASx alpha, PIASX-ALPHA, PIASX-BETA, Protein inhibitor of activated STAT x, protein inhibitor of activated STAT, 2, Protein inhibitor of activated STAT2, SIZ2, zinc finger, MIZ-type containing 4, ZMIZ4 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 9063 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction