missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PI16 (aa 197-285) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (61%), Rat (61%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111189 (PA5-111189. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May inhibit cardiomyocyte growth. [UniProt]
Specifications
Specifications
| Accession Number | Q6UXB8 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 221476 |
| Name | Human PI16 (aa 197-285) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1200009H11Rik; CD364; Cripi; CRISP9; CRISP-9; Cysteine-rich protease inhibitor; cysteine-rich secretory protein 9; microseminoprotein, beta-binding protein; MSMBBP; peptidase inhibitor 16; Pi16; PI-16; protease inhibitor 16; PSEC0164; PSP94-binding protein; PSPBP; UNQ289/PRO328 |
| Common Name | PI16 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction