missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human PI 3-Kinase p55 gamma (NP_003620.3) VersaClone cDNA
Description
Inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
Specifications
Specifications
| Gene | PI 3-Kinase p55 gamma |
| Accession Number | NP_003620.3 |
| Concentration | 10 μg at 0.2 μg/ μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1399 bp |
| Gene ID (Entrez) | 8503 |
| Gene Alias | DKFZp686P05226, FLJ41892, p55, p55-GAMMA, p55PIK, Phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma, phosphatidylinositol 3-kinase regulatory subunit gamma, phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 3 (p55, gamma), phosphoinositide-3-kinase, regulatory subunit 3 (gamma), PI 3 Kinase p55 gamma, PI3K regulatory subunit gamma, PI3-kinase regulatory subunit gamma, PI3-kinase subunit p55-gamma, PIK3R3, PtdIns-3-kinase regulatory subunit gamma, ptdIns-3-kinase regulatory subunit p55-gamma, regulatory subunit, polypeptide 3 (p55, gamma) |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?