missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-WNK1 (T60) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£181.00 - £395.00
Specifications
| Antigen | WNK1 |
|---|---|
| Clone | 1259F |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18736858
![]() |
R&D Systems
MAB4720-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18799777
![]() |
R&D Systems
MAB4720-100 |
100 μg |
£395.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WNK1 Monoclonal specifically detects WNK1 in Human samples. It is validated for Western Blot.Specifications
| WNK1 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| EC 2.7.11, EC 2.7.11.1, Erythrocyte 65 kDa protein, HSAN2hereditary sensory neuropathy, type II, HSN2MGC163339, hWNK1, KDP, KDPMGC163341, KIAA0344, Kinase deficient protein, p65, PHA2C, PRKWNK1, PRKWNK1PSK, prostate-derived sterile 20-like kinase, Protein kinase lysine-deficient 1, Protein kinase with no lysine 1, protein kinase, lysine deficient 1, serine/threonine-protein kinase WNK1, WNK lysine deficient protein kinase 1 | |
| Phosphopeptide containing human WNK1 T60 site Accession # Q9H4A3 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human WNK1 when phosphorylated at T60 in Western blots. |
| 1259F | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9H4A3 | |
| 65125 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title