missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-STAT6 (Y641) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB3717
This item is not returnable.
View return policy
Description
STAT6 Monoclonal specifically detects STAT6 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry.
Specifications
| STAT6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| D12S1644, EC 2.4.1.227, EC 2.7.7.6, IL-4 Stat, IL-4-STAT, signal transducer and activator of transcription 6, signal transducer and activator of transcription 6, interleukin-4 induced, STAT, interleukin-4 induced, STAT, interleukin4-induced, STAT6B, STAT6C, transcription factor IL-4 STAT | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6778 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry | |
| 1248D | |
| Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 2-10 ug/mL | |
| P42226 | |
| STAT6 | |
| Phosphopeptide containing human STAT6 Y641 site. Accession # P42226 | |
| 100 μg | |
| Primary | |
| Detects human STAT6 when phosphorylated at Y641. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction