missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-STAT5a/b (Y694/Y699) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB41901
This item is not returnable.
View return policy
Description
STAT5a/b Monoclonal specifically detects STAT5a/b in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| STAT5a/b | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| MGF, STAT5, STAT5A signal transducer and activator of transcription 5A | |
| Phosphopeptide containing human STAT5b Y699 site (amino acid sequence of this peptide is identical to a corresponding region of human STAT5a containing Y694) Accession # P51692 | |
| 100 μg | |
| Primary | |
| Detects human STAT5a/b when phosphorylated at Y964/Y699 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| 1247C | |
| Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells | |
| P51692 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6776 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction