missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-STAT4 (Y693) Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF4319
This item is not returnable.
View return policy
Description
STAT4 Polyclonal specifically detects STAT4 in Human samples. It is validated for Western Blot.
Specifications
| STAT4 (p tyr693) | |
| Polyclonal | |
| Unconjugated | |
| signal transducer and activator of transcription 4, SLEB11 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6775 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| STAT4 | |
| Phosphopeptide containing human Phospho-STAT4 Y693 site | |
| 100 μg | |
| Primary | |
| Detects human STAT4 when phosphorylated at Y693. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction