missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-PLC-gamma 2 (Y759) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7377
This item is not returnable.
View return policy
Description
PLC-gamma 2 Monoclonal specifically detects PLC-gamma 2 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| PLC-gamma 2 (p tyr759) | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P16885 | |
| PLCG2 | |
| Phosphopeptide containing the human PLC-gamma 2 Y759 site Accession # P16885 | |
| 100 μg | |
| Primary | |
| Detects human Phospho-PLC-gamma 2 (Y759) in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 744757 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.1.4.11, Phosphoinositide phospholipase C-gamma-2, phospholipase C gamma 2, phospholipase C, gamma 2 (phosphatidylinositol-specific), phospholipase C, gamma 2 (phosphatidylyinositol-specific), Phospholipase C-gamma-2,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-2, Phospholipase C-IV, PLCG2, PLCgamma 2, PLC-gamma-2, PLC-IV | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 5336 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction