missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-PGC1 alpha (S571) Antibody, R&D Systems™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF6650
This item is not returnable.
View return policy
Description
PGC1 alpha Polyclonal specifically detects PGC1 alpha in Human samples. It is validated for Western Blot.
Specifications
| PGC1 alpha (p ser571) | |
| Polyclonal | |
| Western Blot 1 ug/mL | |
| PPARGC1A | |
| Phosphopeptide containing the human PGC1 alpha S571 site. | |
| 100 μg | |
| Primary | |
| Detects human PGC1 alpha when phosphorylated at S571 in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| LEM6, Ligand effect modulator 6, ligand effect modulator-6, L-PGC-1alpha, peroxisome proliferative activated receptor, gamma, coactivator 1, alpha, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-3ext, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B4-8a, peroxisome proliferator-activated receptor gamma coactivator 1 alpha transcript variant B5-NT, peroxisome proliferator-activated receptor gamma coactivator 1-alpha, peroxisome proliferator-activated receptor gamma, coactivator 1 alpha, PGC1, PGC-1(alpha), PGC1A, PGC-1-alpha, PGC1APGC-1(alpha), PGC1peroxisome proliferative activated receptor, gamma, coactivator 1, PGC-1v, PPAR gamma coactivator variant form, PPAR gamma coactivator-1, PPARgamma coactivator 1alpha, PPAR-gamma coactivator 1-alpha, PPARGC1, PPARGC1A, PPARGC-1-alpha | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 10891 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction