missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-PDGF R alpha (Y762) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF21141-SP
This item is not returnable.
View return policy
Description
PDGFR alpha Polyclonal specifically detects PDGFR alpha in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PDGFR alpha (p tyr762) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PDGFRA | |
| Phosphopeptide containing human PDGF R alpha Y762 site | |
| 25 μg | |
| Primary | |
| Detects human PDGF R alpha when phosphorylated at Y762. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR alpha, PDGFR2, PDGFRA, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5156 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction