missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-Paxillin (Y31) Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB61641
This item is not returnable.
View return policy
Description
Paxillin Monoclonal specifically detects Paxillin in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Paxillin (p tyr31) | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| FLJ16691, paxillin, PXN | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5829 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| 698239 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PXN | |
| Phosphopeptide containing the human Paxillin Y31 site | |
| 100 μg | |
| Primary | |
| Detects Phospho-Paxillin (Y31) in direct ELISAs and Western blots. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction