missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-Paxillin (Y118) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6164
This item is not returnable.
View return policy
Description
Paxillin Monoclonal specifically detects Paxillin in Human samples. It is validated for Western Blot.
Specifications
| Paxillin (p tyr118) | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ16691, paxillin, PXN | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5829 | |
| Human | |
| Purified |
| Western Blot | |
| 601327 | |
| Western Blot 2 ug/mL | |
| P49023 | |
| PXN | |
| Phosphopeptide containing the human Paxillin Y118 site Accession # P49023 | |
| 100 μg | |
| Primary | |
| Detects Phospho-Paxillin (Y118) in direct ELISAs and Western blots. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction