missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-p53 (S46) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF1489-SP
This item is not returnable.
View return policy
Description
p53 Polyclonal specifically detects p53 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| p53 (p Ser46) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TP53 | |
| Phosphopeptide containing human p53 S46 site | |
| 25 μg | |
| Primary | |
| Detects human p53 when phosphorylated at S46. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Antigen NY-CO-13, BCC7, FLJ92943, LFS1, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, TP53, transformation-related protein 53, TRP53, tumor protein p53, Tumor suppressor p53 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 7157 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction