missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-p53 (S15) Antibody, R&D Systems™
Rabbit Polyclonal Antibody has been used in 7 publications
Marca: R&D Systems AF1043
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
p53 Polyclonal specifically detects p53 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.
Specifica
| p53 (p ser15) | |
| Polyclonal | |
| Unconjugated | |
| Antigen NY-CO-13, BCC7, FLJ92943, LFS1, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, TP53, transformation-related protein 53, TRP53, tumor protein p53, Tumor suppressor p53 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 7157 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 1 ug/500 ug cell lysate, Immunocytochemistry 5-15 ug/mL | |
| TP53 | |
| Phosphopeptide containing human p53 S15 site | |
| 100 μg | |
| Primary | |
| Detects human, monkey, and rat p53 when phosphorylated at S15 and mouse p53 when phosphorylated at S18. | |
| Human | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto