missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Phospho-nNOS (aa 1411-1425) Synthetic Peptide
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Phospho-nNOS (residues 1411-1425) Synthetic Peptide, PEP-188, from Invitrogen.
PEP-188 is a 16 amino acid synthetic peptide. The sequence of this peptide is (amino to carboxy terminus: C N (1411) R L R S E (pS) I A F I E E S K (NH2) (1425).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA1-032. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
Specifications
Specifications
| Gene ID (Entrez) | 4842 |
| Species | Human |
| Purification Method | purified |
| Accession Number | P29475 |
| Common Name | Human Phospho-nNOS (aa 1411-1425) |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | peptide with no preservative |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction