missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Phospho-MSPR/Ron DuoSet IC ELISA
Description
The Human Phospho-MSP R/Ron DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human MSPR/Ron.
Specifications
Specifications
| Product Type | ELISA Development Kit for Phosphorylated Intracellular Proteins |
| Quantity | 1 Kit |
| Species | Human |
| Conjugate | Biotin |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 4486 |
| Synonym | CD136, CD136 antigen, CDw136c-met-related tyrosine kinase, EC 2.7.10, EC 2.7.10.1, macrophage stimulating 1 receptor (c-met-related tyrosine kinase), MSP R, MSP receptor, MST1R, p185-Ron, Protein-tyrosine kinase 8, PTK8, PTK8 protein tyrosine kinase 8, Ron, RONmacrophage-stimulating protein receptor, soluble RON variant 1, soluble RON variant 2, soluble RON variant 3, soluble RON variant 4 |
| Packaging Quantity | 2 Plates |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?