missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-MSK1 (S212) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF1036
This item is not returnable.
View return policy
Description
MSK1/RPS6KA5 Polyclonal specifically detects MSK1/RPS6KA5 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MSK1/RPS6KA5 (p ser212) | |
| Polyclonal | |
| Unconjugated | |
| EC 2.7.11, MGC1911,90 kDa ribosomal protein S6 kinase 5, MSK1EC 2.7.11.1, MSPK1, Nuclear mitogen- and stress-activated protein kinase 1, ribosomal protein S6 kinase alpha-5, ribosomal protein S6 kinase, 90kD, polypeptide 5, ribosomal protein S6 kinase, 90kDa, polypeptide 5, RLPK, RPS6KA5, RSKL, RSK-like protein kinase, S6K-alpha-5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9252 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| RPS6KA5 | |
| Phosphopeptide containing human MSK1 S212 site | |
| 100 μg | |
| Primary | |
| Detects human MSK1 when phosphorylated at S212. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction