missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-MLKL (T357) Antibody, R&D Systems™

Mouse Monoclonal Antibody
£181.00 - £437.00
Specifications
| Antigen | MLKL |
|---|---|
| Clone | 954702 |
| Dilution | Immunohistochemistry 1-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18730148
![]() |
R&D Systems
MAB9187-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18734128
![]() |
R&D Systems
MAB9187-100 |
100 μg |
£437.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MLKL Monoclonal specifically detects MLKL in Human samples. It is validated for Immunohistochemistry.Specifications
| MLKL | |
| Immunohistochemistry 1-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ34389, mixed lineage kinase domain-like, mixed lineage kinase domain-like protein | |
| Phosphopeptide containing the human MLKL T357 site Accession # Q8NB16 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human MLKL when phosphorylated at T357. |
| 954702 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8NB16 | |
| 197259 | |
| IgG2a | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title