missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-LKB1/STK11 (S428) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF5636-SP
This item is not returnable.
View return policy
Description
LKB1/STK11 Polyclonal specifically detects LKB1/STK11 in Human samples. It is validated for Western Blot.
Specifications
| LKB1/STK11 (p ser428) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.11.1, LKB1 serine/threonine kinase 11 (Peutz-Jeghers syndrome), PJS, PJS polarization-related protein LKB1, Renal carcinoma antigen NY-REN-19, serine/threonine kinase 11, serine/threonine-protein kinase 11, Serine/threonine-protein kinase LKB1, STK11 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6794 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q15831 | |
| STK11 | |
| Phosphopeptide containing human LKB1/STK11 S428 site Accession # Q15831 | |
| 25 μg | |
| Primary | |
| Detects human Phospho-LKB1/STK11 (S428) in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction