missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-LAT (Y161) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6334-SP
This item is not returnable.
View return policy
Description
LAT Monoclonal specifically detects LAT in Human samples. It is validated for Western Blot.
Specifications
| LAT (p tyr161) | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| O43561 | |
| LAT | |
| Phosphopeptide containing the human LAT Y161 site Accession # O43561 | |
| 25 μg | |
| Primary | |
| Detects human Phospho-LAT (Y161) in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 627945 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27040 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction