Learn More
Human Phospho-GSK-3 beta (S9) Alexa Fluor 594-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC25062T-100UG
This item is not returnable.
View return policy
Description
GSK-3 beta Monoclonal specifically detects GSK-3 beta in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| GSK-3 beta | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human GSK-3 beta when phosphorylated at S9. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 609739 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK3 beta, GSK-3 beta, GSK3B, GSK3beta isoform | |
| Phosphopeptide containing the human GSK-3 beta S9 site | |
| RUO | |
| 2932 | |
| Human | |
| IgG1 |
Product Content Correction
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction