missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-FAK (Y925) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB44671-SP
This item is not returnable.
View return policy
Description
FAK Monoclonal specifically detects FAK in Human samples. It is validated for Western Blot.
Specifications
| FAK (p tyr925) | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| EC 2.7.10, FADK 1, FADK1, FADKFAK-related non-kinase polypeptide, FAK1FRNK, FAKEC 2.7.10.2, focal adhesion kinase 1, pp125FAK, Protein-tyrosine kinase 2, PTK2, PTK2 protein tyrosine kinase 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5747 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 743234 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PTK2 | |
| Phosphopeptide containing the human FAK Y925 site | |
| 25 μg | |
| Primary | |
| Detects human FAK in direct ELISAs and Western blots. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction