missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-ERK1 (T202/Y204)/ERK2 (T185/Y187) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB1825-SP
This item is not returnable.
View return policy
Description
ERK1/ERK2 Monoclonal specifically detects ERK1/ERK2 in Human samples. It is validated for Western Blot.
Specifications
| ERK1/ERK2 (p Thr202, p Tyr204, p Thr185, p Tyr187) | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects human ERK1 and ERK2 dually phosphorylated at T202/Y204 or T185/Y187, respectively. | |
| Human | |
| Purified |
| Western Blot | |
| 798508 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Phosphopeptide containing the ERK1 T202/Y204 site | |
| 25 μg | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction