missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-ErbB3/Her3 (Y1262) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF5817
This item is not returnable.
View return policy
Description
ErbB3/Her3 Polyclonal specifically detects ErbB3/Her3 in Human samples. It is validated for Western Blot.
Specifications
| ErbB3/Her3 (p tyr1262) | |
| Polyclonal | |
| Unconjugated | |
| c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2065 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| ERBB3 | |
| Phosphopeptide containing human ErbB3 Y1262 site | |
| 100 μg | |
| Primary | |
| Detects human ErbB3 when phosphorylated at Y1262 in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction