missing translation for 'onlineSavingsMsg'
Learn More

Human Phospho-EGFR (Y1086) Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody

£171.00 - £417.00

Specifications

Antigen EGFR
Clone 938835
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 8-25 ug/mL
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18775148
GREENER_CHOICE
View Documents
R&D Systems
MAB89671-SP
25 μg
£171.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18792308
GREENER_CHOICE
View Documents
R&D Systems
MAB89671
100 μg
£417.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

EGFR Monoclonal specifically detects EGFR in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifications

Specifications

EGFR
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 8-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, EGF R, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ErbB, ErbB1, ERBB1PIG61, HER1, HER-1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
EGFR
IgG2a
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
938835
Western Blot, Immunocytochemistry
Unconjugated
Rat
Human
P00533
1956
Phosphopeptide containing the human EGFR Y1086 site Accession # P00533
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human EGFR when phosphorylated at Y1086 in Western blots.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.