missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-CREB (S133) Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB6906
This item is not returnable.
View return policy
Description
CREB Monoclonal specifically detects CREB in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| CREB (p ser133) | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| active transcription factor CREB, cAMP responsive element binding protein 1, cAMP-response element-binding protein-1, cAMP-responsive element-binding protein 1, CREB, CREB1, CREB-1, cyclic AMP-responsive element-binding protein 1, MGC9284, transactivator protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1385 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| 702710 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CREB1 | |
| Phosphopeptide containing the human CREB S133 site | |
| 100 μg | |
| Primary | |
| Detects human CREB when phosphorylated at S133. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction