missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-Chk1 (S345) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF2475
This item is not returnable.
View return policy
Description
Chk1 Polyclonal specifically detects Chk1 in Human samples. It is validated for Western Blot.
Specifications
| Chk1 (p ser345) | |
| Polyclonal | |
| Unconjugated | |
| Checkpoint, S. pombe, homolog of, 1, CHEK1, CHK1 (checkpoint, S.pombe) homolog, CHK1 checkpoint homolog (S. pombe), CHK1serine/threonine-protein kinase Chk1, EC 2.7.11, EC 2.7.11.1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 1111 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| CHEK1 | |
| Phosphopeptide containing human Chk1 S345 site | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat Chk1 when phosphorylated at S345. This antibody does not recognize Chk1 when unphosphorylated at S345. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction