missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-Caveolin-1 (Y14) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7418-SP
This item is not returnable.
View return policy
Description
Caveolin-1 Monoclonal specifically detects Caveolin-1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Caveolin-1 (p tyr14) | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CAV, CAV1, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, Caveolin1, cell growth-inhibiting protein 32, MSTP085, VIP21 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 857 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| 750304 | |
| Western Blot 0.2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q03135 | |
| CAV1 | |
| Phosphopeptide containing the human Caveolin-1 Y14 site Accession # Q03135 | |
| 25 μg | |
| Primary | |
| Detects human Phospho-Caveolin-1 (Y14) in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction