missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Phospho-AMPK alpha 1 (T183) DuoSet IC ELISA

Product Code. 30020968
Click to view available options
Packaging Quantity:
2 Plates
5 Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30020968

Brand: R&D Systems™ DYC35282

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

The Human Phospho-AMPK alpha 1 (T183) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human AMPK alpha 1.
TRUSTED_SUSTAINABILITY

Specifications

Product Type ELISA Development Kit for Phosphorylated Intracellular Proteins
Quantity 1 Kit (for 2 Plates)
Species Human
Conjugate Biotin
Assay Range 62.50 to 4000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
For Use With (Application) ELISA
Gene ID (Entrez) 5562
Synonym AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1,5 '-AMP-activated protein kinase catalytic subunit alpha-1, AMPK, AMPK alpha 1,5 '-AMP-activated protein kinase, catalytic alpha-1 chain, AMPK subunit alpha-1, AMPK1, AMPKa1, EC 2.7.11, EC 2.7.11.1, MGC33776, MGC57364, PRKAA1, protein kinase, AMP-activated, alpha 1 catalytic subunit
Packaging Quantity 2 Plates
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.